siRNA Details
| Gene Symbol: Dcp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133761.3 | NP_598522.3 | 7 | 788 | ||
| GenBank mRNA | AJ344447.1 | - | 7 | 788 | ||
| AK016254.1 | - | 5 | 468 | |||
| AK171707.1 | - | 7 | 712 | |||
| BC066173.1 | - | 7 | 741 | 30049516 | ||

Dcp1a
Genomic Map