siRNA Details
| Gene Symbol: 4930579G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029482.1 | NP_083758.1 | 1 | 114 | ||
| GenBank mRNA | AK016321.1 | - | 1 | 85 | ||
| AK137776.1 | - | 104 | ||||
| BC052483.1 | - | 1 | 95 | |||
| BY309631.1 | - | 1 | 115 | |||

4930579G24Rik
Genomic Map