siRNA Details
| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175229.3 | NP_780438.2 | 10 | 5167 | ||
| GenBank mRNA | AK090123.1 | - | 10 | 5128 | ||
| AK122248.1 | - | 1 | 2352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srrm2| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175229.3 | NP_780438.2 | 10 | 5167 | ||
| GenBank mRNA | AK090123.1 | - | 10 | 5128 | ||
| AK122248.1 | - | 1 | 2352 | |||