siRNA Details
| Gene Symbol: Ms4a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251849.1 | NP_001238778.1 | 2 | 218 | ||
| GenBank mRNA | AK017199.1 | - | 2 | 220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a18| Gene Symbol: Ms4a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251849.1 | NP_001238778.1 | 2 | 218 | ||
| GenBank mRNA | AK017199.1 | - | 2 | 220 | ||