siRNA Details
Gene Symbol: Ms4a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251849.1 | NP_001238778.1 | 7 | 967 | ||
GenBank mRNA | AK017199.1 | - | 7 | 969 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ms4a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251849.1 | NP_001238778.1 | 7 | 967 | ||
GenBank mRNA | AK017199.1 | - | 7 | 969 |