siRNA Details
| Gene Symbol: Zmym2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029498.3 | NP_083774.2 | 2 | 784 | ||
| GenBank mRNA | AK135499.1 | - | 3 | 897 | ||
| BC046507.1 | - | 3 | 830 | |||
| BC141403.1 | - | 2 | 760 | 9056016 | ||
| CA325336.1 | - | 1 | 210 | |||

Zmym2
Genomic Map