siRNA Details
Gene Symbol: Ccdc125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183115.5 | NP_898938.4 | 5, 6 | 889 | ||
GenBank mRNA | BC085249.1 | - | 5, 6 | 607 | 30726150 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183115.5 | NP_898938.4 | 5, 6 | 889 | ||
GenBank mRNA | BC085249.1 | - | 5, 6 | 607 | 30726150 |