siRNA Details
| Gene Symbol: Ncapg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133762.3 | NP_598523.3 | 14 | 1666 | ||
| GenBank mRNA | AY455829.1 | - | 8 | 777 | ||
| BC008539.1 | - | 3 | 347 | |||
| BC076631.1 | - | 13 | 1476 | 30637859 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncapg2| Gene Symbol: Ncapg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133762.3 | NP_598523.3 | 14 | 1666 | ||
| GenBank mRNA | AY455829.1 | - | 8 | 777 | ||
| BC008539.1 | - | 3 | 347 | |||
| BC076631.1 | - | 13 | 1476 | 30637859 | ||