siRNA Details
| Gene Symbol: 5830454E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018010.1 | - | 1 | 323 | ||
| BC100520.1 | - | 1 | 339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830454E08Rik| Gene Symbol: 5830454E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018010.1 | - | 1 | 323 | ||
| BC100520.1 | - | 1 | 339 | |||