siRNA Details
| Gene Symbol: Gpsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029522.2 | NP_083798.2 | 14 | 2291 | ||
| GenBank mRNA | AK018084.1 | - | 4 | 538 | ||
| AK040996.1 | - | 14 | 2180 | |||
| AK143552.1 | - | 14 | 2292 | |||
| AY081187.1 | - | 14 | 2135 | |||
| BC021308.1 | - | 13 | 1846 | 5007832 | ||

Gpsm2
Genomic Map