siRNA Details
| Gene Symbol: Faxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175234.4 | NP_780443.2 | 5, 6 | 1189 | ||
| GenBank mRNA | AK018081.1 | - | 4, 5 | 931 | ||
| AK081616.1 | - | 3, 4 | 766 | |||
| AK144924.1 | - | 5, 6 | 1190 | |||
| BC059864.1 | - | 4, 5 | 479 | |||

Faxc
Genomic Map