siRNA Details
| Gene Symbol: Faxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175234.4 | NP_780443.2 | 6 | 1251 | ||
| GenBank mRNA | AK018081.1 | - | 5 | 993 | ||
| AK081616.1 | - | 4 | 828 | |||
| AK144924.1 | - | 6 | 1252 | |||
| BC059864.1 | - | 5 | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Faxc| Gene Symbol: Faxc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175234.4 | NP_780443.2 | 6 | 1251 | ||
| GenBank mRNA | AK018081.1 | - | 5 | 993 | ||
| AK081616.1 | - | 4 | 828 | |||
| AK144924.1 | - | 6 | 1252 | |||
| BC059864.1 | - | 5 | 541 | |||