siRNA Details
| Gene Symbol: Slc35d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029529.3 | NP_083805.1 | 1 | 515 | ||
| GenBank mRNA | AK042761.1 | - | 1 | 267 | ||
| AK044973.1 | - | 1 | 252 | |||
| AK047610.1 | - | 1 | 516 | |||
| BC139194.1 | - | 1 | 345 | 8862216 | ||

Slc35d3
Genomic Map