siRNA Details
Gene Symbol: Abhd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024465.3 | NP_077785.2 | 8, 9 | 981 | ||
GenBank mRNA | AK018261.1 | - | 8, 9 | 915 | ||
AK160126.1 | - | 8, 9 | 981 | |||
AK178612.1 | - | 1, 2 | 51 | |||
AK185693.1 | - | 1, 2 | 29 | |||
AK198485.1 | - | 1, 2 | 51 | |||
AK202769.1 | - | 1, 2 | 51 | |||
AK216141.1 | - | 1, 2 | 51 | |||
AK217770.1 | - | 1, 2 | 51 | |||
AK219355.1 | - | 2, 3 | 136 | |||
BC002138.1 | - | 8, 9 | 950 | |||
BC002263.1 | - | 4, 5 | 284 | 3495118 |
