siRNA Details
| Gene Symbol: Jakmip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163637.1 | NP_001157109.1 | 15 | 2374 | ||
| GenBank mRNA | AK133750.1 | - | 15 | 2340 | ||
| BC094439.1 | - | 14 | 1936 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jakmip2| Gene Symbol: Jakmip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163637.1 | NP_001157109.1 | 15 | 2374 | ||
| GenBank mRNA | AK133750.1 | - | 15 | 2340 | ||
| BC094439.1 | - | 14 | 1936 | |||