siRNA Details
| Gene Symbol: 0610040J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029554.4 | NP_083830.3 | 4 | 856 | ||
| GenBank mRNA | AK002869.1 | - | 4 | 857 | ||
| AK050792.1 | - | 4 | 988 | |||
| AK159779.1 | - | 4 | 819 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610040J01Rik| Gene Symbol: 0610040J01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029554.4 | NP_083830.3 | 4 | 856 | ||
| GenBank mRNA | AK002869.1 | - | 4 | 857 | ||
| AK050792.1 | - | 4 | 988 | |||
| AK159779.1 | - | 4 | 819 | |||