siRNA Details
| Gene Symbol: Slc18b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183116.2 | NP_898939.2 | 13, 14 | 1460 | ||
| GenBank mRNA | AK003902.1 | - | 1, 2 | 24 | ||
| BC068251.1 | - | 8, 9 | 725 | |||
| BM949736.1 | - | 7, 8 | 644 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc18b1| Gene Symbol: Slc18b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183116.2 | NP_898939.2 | 13, 14 | 1460 | ||
| GenBank mRNA | AK003902.1 | - | 1, 2 | 24 | ||
| BC068251.1 | - | 8, 9 | 725 | |||
| BM949736.1 | - | 7, 8 | 644 | |||