siRNA Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029746.3 | NP_084022.2 | 10 | 1253 | ||
| GenBank mRNA | AK081817.1 | - | 2 | 170 | ||
| AK089920.1 | - | 10 | 1237 | |||
| AK157348.1 | - | 10 | 1215 | |||
| AK169298.1 | - | 10 | 1223 | |||
| AK171726.1 | - | 10 | 1172 | |||
| AK189695.1 | - | 1 | 52 | |||
| BC011435.1 | - | 10 | 1193 | 4010102 | ||
| CX205121.1 | - | 4 | 462 | |||
| W74920.1 | - | 1 | 138 | |||

Cog2
Genomic Map