siRNA Details
| Gene Symbol: Gm1661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145637.1 | NP_001139109.1 | 12 | 1526 | ||
| Gene Symbol: 1700012C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005894.1 | - | 1 | 53 | ||

Gm1661
Genomic Map