siRNA Details
| Gene Symbol: Pifo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200028.1 | NP_001186957.1 | 6 | 751 | ||
| NM_029604.3 | NP_083880.2 | 6 | 703 | |||
| GenBank mRNA | AK006407.1 | - | 6 | 704 | ||
| AW492850.1 | - | 1 | 219 | |||
| AW519725.1 | - | 5 | 415 | |||
| BC049757.1 | - | 6 | 752 | 6773491 | ||
| HM237137.1 | - | 6 | 593 | |||
| HM237138.1 | - | 6 | 751 | |||

Pifo
Genomic Map