siRNA Details
| Gene Symbol: 1700028K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175241.1 | NP_780450.1 | 4 | 428 | ||
| NM_182745.2 | NP_877422.1 | 4 | 428 | |||
| GenBank mRNA | AK006463.1 | - | 4 | 428 | ||
| AK029908.1 | - | 3 | 2057 | |||
| AK032886.1 | - | 5 | 628 | |||
| CN835533.1 | - | 4 | 403 | |||

1700028K03Rik
Genomic Map