siRNA Details
| Gene Symbol: Fam209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029608.1 | NP_083884.1 | 1, 2 | 289 | ||
| GenBank mRNA | AK006503.1 | - | 1, 2 | 296 | ||
| BC137932.1 | - | 1, 2 | 289 | 8860951 | ||
| BC137933.1 | - | 1, 2 | 289 | 8860952 | ||

Fam209
Genomic Map