siRNA Details
| Gene Symbol: Rftn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181397.2 | NP_852062.1 | 4 | 671 | ||
| GenBank mRNA | AK082517.1 | - | 284 | |||
| AK087803.1 | - | 444 | ||||
| AK088466.1 | - | 2 | 208 | |||
| AK152130.1 | - | 4 | 593 | |||
| AK153288.1 | - | 4 | 593 | |||
| AK155870.1 | - | 4 | 635 | |||
| AK170913.1 | - | 4 | 634 | |||
| AK172884.1 | - | 4 | 599 | |||
| BC052074.1 | - | 4 | 671 | 6401192 | ||

Rftn1
Genomic Map