siRNA Details
| Gene Symbol: Ccdc134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172428.2 | NP_766016.2 | 4 | 394 | ||
| GenBank mRNA | AK028514.1 | - | 3 | 520 | ||
| AK049165.1 | - | 4 | 292 | |||
| AK145747.1 | - | 4 | 342 | |||
| AK154557.1 | - | 4 | 398 | |||
| BC072586.1 | - | 4 | 339 | 6828468 | ||
| CJ106860.1 | - | 4 | 395 | |||

Ccdc134
Genomic Map