siRNA Details
| Gene Symbol: 3110002H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029623.2 | NP_083899.1 | 1 | 139 | ||
| GenBank mRNA | AK010309.1 | - | 1 | 133 | ||
| AK144623.1 | - | 1 | 140 | |||
| AK154240.1 | - | 1 | 134 | |||
| AK159691.1 | - | 1 | 134 | |||
| BC012285.1 | - | 1 | 117 | |||
| BC021846.1 | - | 1 | 122 | 5027035 | ||

3110002H16Rik
Genomic Map