siRNA Details
| Gene Symbol: Ly6k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029627.2 | NP_083903.1 | 4 | 473 | ||
| GenBank mRNA | AK010485.1 | - | 4 | 477 | ||
| BC049723.1 | - | 3 | 312 | 6743692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6k| Gene Symbol: Ly6k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029627.2 | NP_083903.1 | 4 | 473 | ||
| GenBank mRNA | AK010485.1 | - | 4 | 477 | ||
| BC049723.1 | - | 3 | 312 | 6743692 | ||