siRNA Details
| Gene Symbol: 1500009C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005178.1 | - | 2 | 333 | ||
| BC023061.1 | - | 2 | 323 | |||
| BC059784.1 | - | 2 | 421 | |||
| BC094356.1 | - | 2 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500009C09Rik| Gene Symbol: 1500009C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005178.1 | - | 2 | 333 | ||
| BC023061.1 | - | 2 | 323 | |||
| BC059784.1 | - | 2 | 421 | |||
| BC094356.1 | - | 2 | 269 | |||