siRNA Details
| Gene Symbol: Lsm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133939.1 | NP_598700.1 | 1, 2 | 135 | ||
| GenBank mRNA | AK088664.1 | - | 1, 2 | 96 | ||
| AK167674.1 | - | 67 | ||||
| BC019458.1 | - | 1, 2 | 135 | 4024697 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm8| Gene Symbol: Lsm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133939.1 | NP_598700.1 | 1, 2 | 135 | ||
| GenBank mRNA | AK088664.1 | - | 1, 2 | 96 | ||
| AK167674.1 | - | 67 | ||||
| BC019458.1 | - | 1, 2 | 135 | 4024697 | ||