siRNA Details
Gene Symbol: Lsm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133939.1 | NP_598700.1 | 4 | 359 | ||
GenBank mRNA | AK008067.1 | - | 4 | 340 | ||
AK088664.1 | - | 4 | 320 | |||
AK154446.1 | - | 1224 | ||||
AK167674.1 | - | 291 | ||||
BC019458.1 | - | 4 | 359 | 4024697 |
