siRNA Details
| Gene Symbol: Prss8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133351.3 | NP_579929.1 | 6 | 1149 | ||
| GenBank mRNA | AB038244.1 | - | 6 | 1130 | ||
| AF188613.1 | - | 6 | 1063 | |||
| AF378085.2 | - | 6 | 1140 | |||
| AK010640.1 | - | 6 | 1031 | |||
| AK078696.1 | - | 6 | 1104 | |||
| AY335911.1 | - | 7 | 997 | |||
| BC003851.1 | - | 6 | 1123 | 3600399 | ||

Prss8
Genomic Map