siRNA Details
| Gene Symbol: Fam101b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029658.1 | NP_083934.1 | 1 | 167 | ||
| GenBank mRNA | AA016881.1 | - | 1 | 56 | ||
| AK171075.1 | - | 1 | 134 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam101b| Gene Symbol: Fam101b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029658.1 | NP_083934.1 | 1 | 167 | ||
| GenBank mRNA | AA016881.1 | - | 1 | 56 | ||
| AK171075.1 | - | 1 | 134 | |||