siRNA Details
| Gene Symbol: 1700034O15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029671.2 | NP_083947.1 | 2 | 329 | ||
| GenBank mRNA | AK006606.1 | - | 2 | 330 | ||
| BC048592.1 | - | 2 | 329 | 6707911 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700034O15Rik| Gene Symbol: 1700034O15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029671.2 | NP_083947.1 | 2 | 329 | ||
| GenBank mRNA | AK006606.1 | - | 2 | 330 | ||
| BC048592.1 | - | 2 | 329 | 6707911 | ||