siRNA Details
| Gene Symbol: 1700123K08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029693.2 | NP_083969.1 | 1 | 92 | ||
| GenBank mRNA | AK007254.1 | - | 1 | 93 | ||
| BC147525.1 | - | 1 | 80 | 9056517 | ||
| BC147536.1 | - | 1 | 80 | 9056527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700123K08Rik| Gene Symbol: 1700123K08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029693.2 | NP_083969.1 | 1 | 92 | ||
| GenBank mRNA | AK007254.1 | - | 1 | 93 | ||
| BC147525.1 | - | 1 | 80 | 9056517 | ||
| BC147536.1 | - | 1 | 80 | 9056527 | ||