siRNA Details
| Gene Symbol: Cfap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163638.1 | NP_001157110.1 | 3 | 293 | ||
| NM_001163639.1 | NP_001157111.1 | 3 | 293 | |||
| NM_029698.1 | NP_083974.1 | 3 | 231 | |||
| GenBank mRNA | AK017232.1 | - | 3 | 294 | ||
| BC138269.1 | - | 3 | 290 | 8861288 | ||
| BC145225.1 | - | 3 | 290 | 9053777 | ||
| BC145226.1 | - | 3 | 290 | 9053778 | ||

Cfap70
Genomic Map