siRNA Details
| Gene Symbol: Cfap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163638.1 | NP_001157110.1 | 22 | 2708 | ||
| NM_001163639.1 | NP_001157111.1 | 21 | 2627 | |||
| NM_029698.1 | NP_083974.1 | 22 | 2697 | |||
| GenBank mRNA | AK029536.1 | - | 22 | 2698 | ||
| BC138269.1 | - | 21 | 2624 | 8861288 | ||
| BC145225.1 | - | 22 | 2705 | 9053777 | ||
| BC145226.1 | - | 21 | 2624 | 9053778 | ||

Cfap70
Genomic Map