siRNA Details
| Gene Symbol: Ctrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033875.2 | NP_001029047.1 | 7 | 813 | ||
| GenBank mRNA | AK007772.1 | - | 6 | 641 | ||
| BC103715.1 | - | 7 | 811 | 6430373 | ||
| BC115516.1 | - | 7 | 813 | 40101127 | ||
| BC115517.1 | - | 7 | 813 | 40101128 | ||

Ctrc
Genomic Map