siRNA Details
| Gene Symbol: Arpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029711.1 | NP_083987.1 | 8, 9 | 1063 | ||
| GenBank mRNA | AK008777.1 | - | 4, 5 | 509 | ||
| AK083263.1 | - | 8, 9 | 873 | |||
| AK151513.1 | - | 8, 9 | 1037 | |||
| AK152356.1 | - | 9, 10 | 888 | |||
| AK170061.1 | - | 9, 10 | 862 | |||
| BC099504.1 | - | 7, 8 | 715 | |||
| BC115750.1 | - | 8, 9 | 861 | 40051600 | ||

Arpc2
Genomic Map