siRNA Details
| Gene Symbol: Arpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029711.1 | NP_083987.1 | 7 | 896 | ||
| GenBank mRNA | AK008777.1 | - | 3 | 342 | ||
| AK083263.1 | - | 7 | 706 | |||
| AK151513.1 | - | 7 | 870 | |||
| AK152356.1 | - | 8 | 721 | |||
| AK170061.1 | - | 8 | 695 | |||
| BC099504.1 | - | 6 | 548 | |||
| BC115750.1 | - | 7 | 694 | 40051600 | ||

Arpc2
Genomic Map