siRNA Details
| Gene Symbol: Trdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029726.2 | NP_084002.2 | 10 | 1130 | ||
| GenBank mRNA | AF223417.1 | - | 9 | 992 | ||
| BC034343.1 | - | 9 | 992 | 3589077 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trdn| Gene Symbol: Trdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029726.2 | NP_084002.2 | 10 | 1130 | ||
| GenBank mRNA | AF223417.1 | - | 9 | 992 | ||
| BC034343.1 | - | 9 | 992 | 3589077 | ||