siRNA Details
| Gene Symbol: Gsdma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029727.2 | NP_084003.2 | 11 | 1315 | ||
| GenBank mRNA | AK008613.1 | - | 9 | 968 | ||
| AK008663.1 | - | 11 | 1311 | |||
| BC109335.1 | - | 10 | 1218 | 40052139 | ||
| BC109336.1 | - | 11 | 1249 | 40052146 | ||

Gsdma2
Genomic Map