siRNA Details
| Gene Symbol: Gsdma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029727.2 | NP_084003.2 | 4 | 422 | ||
| GenBank mRNA | AK008663.1 | - | 4 | 419 | ||
| AK008685.1 | - | 5 | 454 | |||
| BC109335.1 | - | 3 | 325 | 40052139 | ||
| BC109336.1 | - | 4 | 356 | 40052146 | ||

Gsdma2
Genomic Map