siRNA Details
| Gene Symbol: Mettl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176917.4 | NP_795891.2 | 4 | 1259 | ||
| GenBank mRNA | AK157268.1 | - | 4 | 1260 | ||
| BC099943.1 | - | 3 | 1676 | 30020588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl4| Gene Symbol: Mettl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176917.4 | NP_795891.2 | 4 | 1259 | ||
| GenBank mRNA | AK157268.1 | - | 4 | 1260 | ||
| BC099943.1 | - | 3 | 1676 | 30020588 | ||