siRNA Details
| Gene Symbol: Armc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133972.2 | NP_598733.2 | 4 | 772 | ||
| GenBank mRNA | AK010815.1 | - | 4 | 800 | ||
| AK049351.1 | - | 4 | 488 | |||
| AK080609.1 | - | 4 | 477 | |||
| AK081550.1 | - | 4 | 817 | |||
| BC043070.1 | - | 4 | 772 | 6403587 | ||
| BC048396.1 | - | 4 | 733 | |||
| BC055779.1 | - | 4 | 772 | 6403587 | ||

Armc6
Genomic Map