siRNA Details
| Gene Symbol: Nubpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029760.2 | NP_084036.2 | 1, 2 | 136 | ||
| GenBank mRNA | AK010827.1 | - | 1, 2 | 118 | ||
| AK083643.1 | - | 1, 2 | 126 | |||
| AK141257.1 | - | 1, 2 | 100 | |||
| AK152377.1 | - | 1, 2 | 137 | |||
| BC062175.1 | - | 1, 2 | 117 | 30291107 | ||

Nubpl
Genomic Map