siRNA Details
| Gene Symbol: Spopl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165997.1 | NP_001159469.1 | 10 | 1382 | ||
| NM_001165998.1 | NP_001159470.1 | 9 | 1246 | |||
| NM_029773.2 | NP_084049.2 | 9 | 1285 | |||
| GenBank mRNA | AK014913.1 | - | 4 | 343 | ||
| AK162056.1 | - | 9 | 1100 | |||
| BC111867.1 | - | 10 | 1256 | 40056321 | ||
| CD541905.1 | - | 4 | 379 | |||

Spopl
Genomic Map