siRNA Details
| Gene Symbol: Lrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029796.2 | NP_084072.1 | 2 | 165 | ||
| GenBank mRNA | AB055885.1 | - | 2 | 125 | ||
| AF403429.1 | - | 2 | 108 | |||
| AK004940.1 | - | 1 | 113 | |||
| AK009555.1 | - | 1 | 121 | |||
| BC030733.1 | - | 2 | 165 | 5123909 | ||
| BC092077.1 | - | 2 | 110 | 4975749 | ||

Lrg1
Genomic Map