siRNA Details
| Gene Symbol: Mnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029797.3 | NP_084073.2 | 5 | 444 | ||
| GenBank mRNA | AK011664.1 | - | 5 | 354 | ||
| BC027741.1 | - | 5 | 371 | 5035682 | ||
| BG070803.2 | - | 4 | 427 | |||
| BY710901.1 | - | 5 | 354 | |||
| Gene Symbol: Mnd1-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK156325.1 | - | 1 | 2144 | ||

Mnd1
Genomic Map