siRNA Details
| Gene Symbol: 2810429I04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034854.1 | - | 1 | 1317 | ||
| AK043092.1 | - | 2 | 169 | |||
| AK043278.1 | - | 2 | 88 | |||
| AK084238.1 | - | 1 | 1327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810429I04Rik| Gene Symbol: 2810429I04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034854.1 | - | 1 | 1317 | ||
| AK043092.1 | - | 2 | 169 | |||
| AK043278.1 | - | 2 | 88 | |||
| AK084238.1 | - | 1 | 1327 | |||