siRNA Details
| Gene Symbol: St5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001326.1 | NP_001001326.1 | 13, 14 | 2671 | ||
| NM_029811.2 | NP_084087.2 | 12, 13 | 1420 | |||
| GenBank mRNA | AK008100.1 | - | 5, 6 | 494 | ||
| BC030391.1 | - | 9, 10 | 1146 | 4950637 | ||
| BC032266.1 | - | 5, 6 | 566 | |||
| BC051034.1 | - | 13, 14 | 2852 | 5345299 | ||
| CF749727.1 | - | 6, 7 | 596 | |||

St5
Genomic Map