siRNA Details
| Gene Symbol: St5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001326.1 | NP_001001326.1 | 17, 18 | 3152 | ||
| NM_029811.2 | NP_084087.2 | 16, 17 | 1901 | |||
| GenBank mRNA | AK008100.1 | - | 9, 10 | 975 | ||
| BC030391.1 | - | 13, 14 | 1627 | 4950637 | ||
| BC032266.1 | - | 9, 10 | 1047 | |||
| BC051034.1 | - | 17, 18 | 3333 | 5345299 | ||

St5
Genomic Map